This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DCDC2C SIRNA Assays ›
Gene Symbol:
DCDC2C
Gene Name:
doublecortin domain containing 2C
Gene Aliases:
-
Chromosome Location:
Chr.2: 3703592 - 3847411 on Build GRCh38
UniGene ID:
Hs.304233
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS157850
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DCDC2C
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287444.1 NP_001274373.1 1, 2 436
XM_006711894.3 XP_006711957.1 1, 2 430
XM_011510393.2 XP_011508695.1 1, 2 430
XM_011510394.2 XP_011508696.1 1, 2 430
XM_017004836.1 XP_016860325.1 1, 2 430
XM_017004837.1 XP_016860326.1 1, 2 430
XM_017004838.1 XP_016860327.1 1, 2 430
GenBank mRNA DW010024.1 - 1, 2 131
HY010234.1 - 1, 2 436

Back To Top

More Information


Gene Ontology Categories:

Function(s)

intracellular signal transduction

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline