siRNA Details
| Gene Symbol: VGLL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320493.1 | NP_001307422.1 | 1 | 420 | ||
| NM_001320494.1 | NP_001307423.1 | 1 | 420 | |||
| NM_016206.3 | NP_057290.2 | 1 | 420 | |||
| XM_006713138.3 | XP_006713201.1 | 1 | 590 | |||
| GenBank mRNA | BC094780.1 | - | 1 | 416 | 30528902 | |
| BX391744.2 | - | 3 | 759 | |||
| CF995043.1 | - | 443 | ||||
| CR936700.1 | - | 1 | 57 | |||

VGLL3
Genomic Map