siRNA Details
| Gene Symbol: LOC102724708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136223.1 | 1 | 411 | |||
| NR_136224.1 | 1 | 411 | ||||
| GenBank mRNA | BC030256.1 | - | 1 | 411 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724708| Gene Symbol: LOC102724708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136223.1 | 1 | 411 | |||
| NR_136224.1 | 1 | 411 | ||||
| GenBank mRNA | BC030256.1 | - | 1 | 411 | ||