siRNA Details
| Gene Symbol: LOC101928443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120529.1 | 1 | 798 | |||
| GenBank mRNA | BC039105.1 | - | 1 | 798 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928443| Gene Symbol: LOC101928443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120529.1 | 1 | 798 | |||
| GenBank mRNA | BC039105.1 | - | 1 | 798 | ||