This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PRRT4 SIRNA Assays ›
Gene Symbol:
PRRT4
Gene Name:
proline rich transmembrane protein 4
Gene Aliases:
-
Chromosome Location:
Chr.7: 128350325 - 128362067 on Build GRCh38
UniGene ID:
Hs.381096
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS161300
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PRRT4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114726.2 NP_001108198.2 2 420
NM_001174164.1 NP_001167635.1 3 537
XM_005250342.3 XP_005250399.1 2 537
XM_005250343.3 XP_005250400.1 2 446
XM_006715985.3 XP_006716048.1 2 550
XM_017012205.1 XP_016867694.1 3 1356
XM_017012206.1 XP_016867695.1 2 1329
GenBank mRNA BX355277.2 - 3 483

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-stzfq:80/100.66.77.128:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0