This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GPR107 SIRNA Assays ›
Gene Symbol:
GPR107
Gene Name:
G protein-coupled receptor 107
Gene Aliases:
GCDRP, LUSTR1, bA138E2.2
Chromosome Location:
Chr.9: 130053706 - 130140169 on Build GRCh38
UniGene ID:
Hs.512461
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS164968
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GPR107
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136557.1 NP_001130029.1 20 6221
NM_001136558.1 NP_001130030.1 19 6134
NM_020960.4 NP_066011.2 18 6077
GenBank mRNA AB046844.1 - 20 5992
AJ420422.1 - 1 609
AK000825.1 - 1 605
AK021506.1 - 1 1614
BC007544.1 - 1 820

Back To Top

More Information


Set Membership:

G-protein coupled receptor

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

clathrin-mediated endocytosis
clathrin heavy chain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nspbl:80/100.66.74.46:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline