This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SPANXN1 SIRNA Assays ›
Gene Symbol:
SPANXN1 SPANXN2
Gene Name:
SPANX family member N1
SPANX family member N2
Gene Aliases:
CT11.6
CT11.7, SPANX-N2
Chromosome Location:
Chr.X: 145247587 - 145256208 on Build GRCh38
Chromosome Location:
Chr.X: 143712035 - 143720668 on Build GRCh38
UniGene ID:
Hs.551270
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID HSS166733
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SPANXN1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009614.2 NP_001009614.1 1 13
GenBank mRNA DQ336118.1 - 1 13
DQ336119.1 - 1 13
DQ336120.1 - 1 13
DQ336121.1 - 1 13
Gene Symbol: SPANXN2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA DQ336111.1 - 1 13

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-5d9f4b9bfc-mr75l:80/100.66.78.52:80.
git-commit: 52aea788d112809596cd23e639b0bc7998ca5a01
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-2025.11.48-1.0