This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF704 SIRNA Assays ›
Gene Symbol:
ZNF704
Gene Name:
zinc finger protein 704
Gene Aliases:
Gig1
Chromosome Location:
Chr.8: 80628451 - 80884416 on Build GRCh38
UniGene ID:
Hs.434957
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS166907
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF704
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033723.2 NP_001028895.1 2, 3 455
XM_005251279.4 XP_005251336.2 2, 3 1086
XM_017013725.1 XP_016869214.1 2, 3 502
GenBank mRNA AK131274.1 - 2, 3 455
BC136844.1 - 2, 3 268 9020835
BC136845.1 - 2, 3 268 9020836

Back To Top

More Information


Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Process(es)

metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline