This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C2ORF82 SIRNA Assays ›
Gene Symbol:
C2orf82
Gene Name:
chromosome 2 open reading frame 82
Gene Aliases:
UNQ830
Chromosome Location:
Chr.2: 232865678 - 232878700 on Build GRCh38
UniGene ID:
Hs.657509
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS167449
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C2orf82
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206895.1 NP_996778.1 1, 2 125
XM_017004080.1 XP_016859569.1 2, 3 1155
XM_017004081.1 XP_016859570.1 2, 3 893
XM_017004082.1 XP_016859571.1 3, 4 1213
XM_017004083.1 XP_016859572.1 2, 3 925
XM_017004084.1 XP_016859573.1 2, 3 188
GenBank mRNA AY358535.1 - 1, 2 125
BC130307.1 - 1, 2 123 40146338
BC130309.1 - 1, 2 123 40146340

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fh6lz:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline