siRNA Details
| Gene Symbol: LOC51145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122032.1 | 2 | 587 | |||
| GenBank mRNA | AA421133.1 | - | 1 | 122 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC51145| Gene Symbol: LOC51145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122032.1 | 2 | 587 | |||
| GenBank mRNA | AA421133.1 | - | 1 | 122 | ||