siRNA Details
| Gene Symbol: ZNRD1ASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC067890.1 | - | 6 | 1703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNRD1ASP| Gene Symbol: ZNRD1ASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC067890.1 | - | 6 | 1703 | ||