siRNA Details
| Gene Symbol: VAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006373.3 | NP_006364.2 | 1 | 300 | ||
| GenBank mRNA | AK290460.1 | - | 1 | 276 | ||
| BC001913.1 | - | 1 | 169 | |||
| BC008725.2 | - | 1 | 224 | 3162543 | ||
| BC014279.2 | - | 1 | 206 | 3834206 | ||
| BC015041.2 | - | 1 | 238 | 3919901 | ||
| BC090858.1 | - | 1 | 238 | 6528247 | ||
| BT007369.1 | - | 1 | 180 | |||

VAT1
Genomic Map