This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CHN2 SIRNA Assays ›
Gene Symbol:
CHN2
Gene Name:
chimerin 2
Gene Aliases:
ARHGAP3, BCH, CHN2-3, RHOGAP3
Chromosome Location:
Chr.7: 29146547 - 29514335 on Build GRCh38
UniGene ID:
Hs.654611
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS174195
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CHN2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039936.2 NP_001035025.1 7 1326
NM_001293069.1 NP_001279998.1 14 1597
NM_001293070.1 NP_001279999.1 14 1811
NM_001293071.1 NP_001280000.1 12 1716
NM_001293072.1 NP_001280001.1 13 1529
NM_001293073.1 NP_001280002.1 7 1191
NM_001293075.1 NP_001280004.1 6 1053
NM_001293076.1 NP_001280005.1 6 1152
NM_001293077.1 NP_001280006.1 5 1110
NM_001293078.1 NP_001280007.1 5 1014
NM_001293079.1 NP_001280008.1 5 1082
NM_001293080.1 NP_001280009.1 6 1188
NM_001293081.1 NP_001280010.1 7 841
NM_004067.3 NP_004058.1 13 1772
NR_120524.1 4 862
NR_120525.1 5 932
XM_011515105.2 XP_011513407.1 16 1652
XM_011515106.2 XP_011513408.1 15 1613
XM_011515107.2 XP_011513409.1 15 2395
XM_017011721.1 XP_016867210.1 17 1670
XM_017011722.1 XP_016867211.1 16 2413
GenBank mRNA AK026415.1 - 7 1326
AK126784.1 - 2 2530
AK293959.1 - 14 1391
AK294032.1 - 13 1529
AK294216.1 - 7 841
AK294271.1 - 14 1598
AK301235.1 - 7 850
AK313021.1 - 13 1546
AK316030.1 - 7 850
BC112155.1 - 13 1517 8327623
EU732750.1 - 4 463
EU732751.1 - 5 533
EU732752.1 - 7 792
EU732753.1 - 6 654
EU732754.1 - 6 753
EU732755.1 - 5 615
EU732756.1 - 5 711
EU732757.1 - 5 683
EU732758.1 - 6 789
EU732759.1 - 4 401
EU732760.1 - 12 1279
EU732761.1 - 4 244
EU732762.1 - 10 903
EU732763.1 - 6 303
EU732764.1 - 7 507
EU732765.1 - 8 477
EU732766.1 - 8 653
GQ924106.1 - 14 1598
U28926.1 - 13 1569

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of signal transduction
intracellular signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
SH3/SH2 adaptor activity
GTPase activator activity
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline