This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KLHL23 SIRNA Assays ›
Gene Symbol:
KLHL23 PHOSPHO2-KLHL23
Gene Name:
kelch like family member 23
PHOSPHO2-KLHL23 readthrough
Gene Aliases:
DITHP
-
Chromosome Location:
Chr.2: 169733846 - 169751886 on Build GRCh38
Chromosome Location:
Chr.2: 169694454 - 169751886 on Build GRCh38
UniGene ID:
Hs.655150
Hs.655150
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID HSS175667
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KLHL23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144711.5 NP_653312.2 2 1059
GenBank mRNA AK095131.1 - 635
BC010437.1 - 893 3354344
Gene Symbol: PHOSPHO2-KLHL23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199290.1 NP_001186219.1 4 1190
GenBank mRNA AK090653.1 - 4 1153
AX746559.1 - 4 1153

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
ubiquitin-protein transferase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0