This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DGKQ SIRNA Assays ›
Gene Symbol:
DGKQ
Gene Name:
diacylglycerol kinase theta
Gene Aliases:
DAGK, DAGK4, DAGK7
Chromosome Location:
Chr.4: 958884 - 973560 on Build GRCh38
UniGene ID:
Hs.584858
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS175921
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DGKQ
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001347.3 NP_001338.2 19, 20 2391
XM_011513411.1 XP_011511713.1 19, 20 2376
GenBank mRNA AB209503.1 - 19, 20 2142
BC063801.1 - 19, 20 2399 6526640
GU727647.1 - 19, 20 2329
L38707.1 - 19, 20 2351

Back To Top

More Information


Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
G-protein coupled receptor signaling pathway
protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
cAMP-mediated signaling
platelet activation
response to ATP
thrombin receptor signaling pathway
protein kinase C signaling
diacylglycerol kinase activity
protein binding
ATP binding
kinase binding
activating transcription factor binding
phospholipase binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline