This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF383 SIRNA Assays ›
Gene Symbol:
ZNF383
Gene Name:
zinc finger protein 383
Gene Aliases:
HSD17
Chromosome Location:
Chr.19: 37217243 - 37248740 on Build GRCh38
UniGene ID:
Hs.590975
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS175982
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF383
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152604.1 NP_689817.1 5 1399
XM_005258585.2 XP_005258642.1 6 1657
XM_005258587.2 XP_005258644.1 5 1534
XM_005258588.2 XP_005258645.1 6 1296
XM_011526586.2 XP_011524888.1 7 2402
XM_011526587.2 XP_011524889.1 6 2369
XM_011526588.2 XP_011524890.1 5 2551
XM_011526589.2 XP_011524891.1 5 1232
XM_011526590.2 XP_011524892.1 4 1197
XM_017026422.1 XP_016881911.1 7 1808
XM_017026423.1 XP_016881912.1 7 2402
XM_017026424.1 XP_016881913.1 5 1448
GenBank mRNA AK091760.1 - 5 1960
AK093182.1 - 5 1399
AK122892.1 - 8 1725
AY260141.1 - 4 1194
AY438646.1 - 5 1399

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0