This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ESRRA SIRNA Assays ›
Gene Symbol:
ESRRA
Gene Name:
estrogen related receptor alpha
Gene Aliases:
ERR1, ERRa, ERRalpha, ESRL1, NR3B1
Chromosome Location:
Chr.11: 64305528 - 64316743 on Build GRCh38
UniGene ID:
Hs.110849
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS176623
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ESRRA
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282450.1 NP_001269379.1 6 1209
NM_001282451.1 NP_001269380.1 6 1193
NM_004451.4 NP_004442.3 6 1196
XM_017017313.1 XP_016872802.1 5 1700
GenBank mRNA BC007915.2 - 5 666
BC033701.1 - 4 605
BC063795.1 - 6 1146 6527166
BC092470.1 - 6 1209 30336569
BC093720.1 - 5 997 7939565
BC093722.1 - 5 997 7939567
HQ692867.1 - 5 975
L38487.1 - 6 1149
X51416.1 - 1329

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
mitochondrion organization
intracellular receptor signaling pathway
response to estradiol
regulation of cell proliferation
steroid hormone mediated signaling pathway
regulation of osteoblast differentiation
regulation of osteoclast differentiation
positive regulation of transcription from RNA polymerase II promoter
cartilage development
positive regulation of cellular response to insulin stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
steroid binding
protein binding
zinc ion binding
protein domain specific binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nzkhn:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline