This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other EVX1 SIRNA Assays ›
Gene Symbol:
EVX1
Gene Name:
even-skipped homeobox 1
Gene Aliases:
EVX-1
Chromosome Location:
Chr.7: 27242545 - 27247819 on Build GRCh38
UniGene ID:
Hs.369879
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS176635
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: EVX1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304519.1 NP_001291448.1 2 1042
NM_001304520.1 NP_001291449.1 3 1094
NM_001989.4 NP_001980.1 2 1097
GenBank mRNA AK298822.1 - 3 271
AK314155.1 - 2 887
X60655.1 - 2 839

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

embryo development ending in birth or egg hatching
regulation of transcription from RNA polymerase II promoter involved in ventral spinal cord interneuron specification
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-tmv5f:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline