This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM26 SIRNA Assays ›
Gene Symbol:
TMEM26
Gene Name:
transmembrane protein 26
Gene Aliases:
-
Chromosome Location:
Chr.10: 61406642 - 61453450 on Build GRCh38
UniGene ID:
Hs.623955
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS176718
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM26
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178505.7 NP_848600.2 4 891
NR_134507.1 4 891
NR_134508.1 4 891
XM_011539450.2 XP_011537752.1 4 679
XM_011539451.1 XP_011537753.1 4 679
XM_017015829.1 XP_016871318.1 4 679
GenBank mRNA AK125611.1 - 4 679
BC042872.1 - 4 608 5164050
CR749606.1 - 4 891

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-7458x:80/100.66.72.156:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline