siRNA Details
| Gene Symbol: OR6Q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005186.2 | NP_001005186.2 | 1 | 456 | ||
| GenBank mRNA | BC151146.1 | - | 1 | 479 | 9021017 | |
| BC151147.1 | - | 1 | 479 | 9021018 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR6Q1| Gene Symbol: OR6Q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005186.2 | NP_001005186.2 | 1 | 456 | ||
| GenBank mRNA | BC151146.1 | - | 1 | 479 | 9021017 | |
| BC151147.1 | - | 1 | 479 | 9021018 | ||