siRNA Details
| Gene Symbol: ANGPTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178127.4 | NP_835228.2 | 5, 6 | 1039 | ||
| GenBank mRNA | AK291523.1 | - | 4, 5 | 503 | ||
| AY169281.1 | - | 4, 5 | 497 | |||
| AY358827.1 | - | 4, 5 | 487 | |||
| BC049170.1 | - | 5, 6 | 1042 | 5285447 | ||
| BC053957.1 | - | 5, 6 | 1042 | |||

ANGPTL5
Genomic Map