This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SYCE2 SIRNA Assays ›
Gene Symbol:
SYCE2
Gene Name:
synaptonemal complex central element protein 2
Gene Aliases:
CESC1
Chromosome Location:
Chr.19: 12898229 - 12919674 on Build GRCh38
UniGene ID:
Hs.655173
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS177769
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SYCE2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105578.1 NP_001099048.1 5 607
XM_005259848.4 XP_005259905.1 5 679
XM_011527882.2 XP_011526184.1 5 1006
XM_011527883.2 XP_011526185.1 5 1006
GenBank mRNA AK097443.1 - 4 1857
AK302373.1 - 5 597
BC035819.2 - 4 576

Back To Top

More Information


Gene Ontology Categories:

Function(s)

synaptonemal complex assembly
cell division

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-ftbch:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0