This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF658 SIRNA Assays ›
Gene Symbol:
ZNF658
Gene Name:
zinc finger protein 658
Gene Aliases:
-
Chromosome Location:
Chr.9: 66900725 - 66932141 on Build GRCh38
UniGene ID:
Hs.522147
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS178070
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF658
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317916.1 NP_001304845.1 4, 5 542
NM_033160.6 NP_149350.3 4, 5 385
NR_134255.1 3, 4 430
XM_005272515.4 XP_005272572.1 4, 5 492
XM_011545679.2 XP_011543981.1 4, 5 316
XM_017014614.1 XP_016870103.1 3, 4 2678
GenBank mRNA DA120232.1 - 4, 5 324

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
ribosome biogenesis
negative regulation of transcription, DNA-templated
cellular response to zinc ion
transcription regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline