siRNA Details
| Gene Symbol: KRTAP7-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181606.2 | NP_853637.2 | 1 | 240 | ||
| GenBank mRNA | AB096962.1 | - | 1 | 240 | ||
| AJ457063.1 | - | 1 | 117 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP7-1| Gene Symbol: KRTAP7-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181606.2 | NP_853637.2 | 1 | 240 | ||
| GenBank mRNA | AB096962.1 | - | 1 | 240 | ||
| AJ457063.1 | - | 1 | 117 | |||