siRNA Details
| Gene Symbol: SLC35D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008783.2 | NP_001008783.1 | 2 | 1394 | ||
| GenBank mRNA | AK295424.1 | - | 2 | 1243 | ||
| AK316095.1 | - | 2 | 1394 | |||
| BC067217.1 | - | 1 | 627 | |||
| BC087842.1 | - | 2 | 1371 | 6191889 | ||

SLC35D3
Genomic Map