This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF445 SIRNA Assays ›
Gene Symbol:
ZNF445
Gene Name:
zinc finger protein 445
Gene Aliases:
ZKSCAN15, ZNF168, ZSCAN47
Chromosome Location:
Chr.3: 44439770 - 44477670 on Build GRCh38
UniGene ID:
Hs.250481
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS179858
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF445
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181489.5 NP_852466.1 8 1530
XM_005265102.2 XP_005265159.1 7 1388
XM_005265103.3 XP_005265160.1 5 660
XM_011533674.2 XP_011531976.1 7 1353
XM_011533675.2 XP_011531977.1 5 624
XM_017006345.1 XP_016861834.1 7 1409
XM_017006346.1 XP_016861835.1 7 1352
GenBank mRNA AY262260.1 - 6 1333
AY295873.2 - 8 1530
BC101486.1 - 6 1244 8068992
BC143439.1 - 6 1209 9051947

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rrb5s:80/100.66.77.251:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline