This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C16ORF86 SIRNA Assays ›
Gene Symbol:
C16orf86
Gene Name:
chromosome 16 open reading frame 86
Gene Aliases:
-
Chromosome Location:
Chr.16: 67666806 - 67670523 on Build GRCh38
UniGene ID:
Hs.632208
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS180294
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C16orf86
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012984.2 NP_001013002.2 3 590
XM_005255952.4 XP_005256009.1 3 598
GenBank mRNA AK123796.1 - 2 940
BC032064.1 - 3 567
BC064498.1 - 3 636
BC082249.1 - 3 636
BC132878.1 - 1 505
BC132880.1 - 1 505

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline