siRNA Details
| Gene Symbol: KRT82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033033.3 | NP_149022.3 | 9 | 1543 | ||
| GenBank mRNA | AJ628419.1 | - | 9 | 1506 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT82| Gene Symbol: KRT82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033033.3 | NP_149022.3 | 9 | 1543 | ||
| GenBank mRNA | AJ628419.1 | - | 9 | 1506 | ||