This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF772 SIRNA Assays ›
Gene Symbol:
ZNF772
Gene Name:
zinc finger protein 772
Gene Aliases:
-
Chromosome Location:
Chr.19: 57464198 - 57480950 on Build GRCh38
UniGene ID:
Hs.388810
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS180757
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF772
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024596.2 NP_001019767.1 2 318
NM_001144068.1 NP_001137540.1 2 318
XM_005258943.4 XP_005259000.1 2 3698
XM_011526980.2 XP_011525282.1 2 3698
XM_017026813.1 XP_016882302.1 2 3698
GenBank mRNA AK294941.1 - 2 273
AK302574.1 - 2 280
BX647068.1 - 2 318

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fbxsq:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline