siRNA Details
| Gene Symbol: LINC01144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024270.1 | 1 | 386 | |||
| GenBank mRNA | BC006119.1 | - | 1 | 105 | ||
| BC035497.2 | - | 1 | 386 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01144| Gene Symbol: LINC01144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024270.1 | 1 | 386 | |||
| GenBank mRNA | BC006119.1 | - | 1 | 105 | ||
| BC035497.2 | - | 1 | 386 | |||