siRNA Details
| Gene Symbol: FAM183A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101376.2 | NP_001094846.2 | 1 | 86 | ||
| XM_005270875.4 | XP_005270932.1 | 1 | 915 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM183A| Gene Symbol: FAM183A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101376.2 | NP_001094846.2 | 1 | 86 | ||
| XM_005270875.4 | XP_005270932.1 | 1 | 915 | |||