siRNA Details
| Gene Symbol: MYL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002477.1 | NP_002468.1 | 5 | 422 | ||
| XM_006713886.2 | XP_006713949.1 | 5 | 799 | |||
| XM_017008245.1 | XP_016863734.1 | 6 | 2100 | |||
| XM_017008246.1 | XP_016863735.1 | 7 | 1969 | |||
| XM_017008247.1 | XP_016863736.1 | 5 | 1559 | |||
| XM_017008248.1 | XP_016863737.1 | 6 | 582 | |||
| GenBank mRNA | AK123974.1 | - | 6 | 2718 | ||
| AK125529.1 | - | 6 | 1542 | |||
| BC040050.1 | - | 5 | 542 | 6014371 | ||
| L03785.1 | - | 5 | 422 | |||

MYL5
Genomic Map