This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BOLA1 SIRNA Assays ›
Gene Symbol:
BOLA1
Gene Name:
bolA family member 1
Gene Aliases:
CGI-143
Chromosome Location:
Chr.1: 149899582 - 149900795 on Build GRCh38
UniGene ID:
Hs.13880
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS181813
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BOLA1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321025.1 NP_001307954.1 2 497
NM_001321026.1 NP_001307955.1 2 555
NM_016074.4 NP_057158.1 2 330
XM_006711348.3 XP_006711411.1 2 363
GenBank mRNA AF151901.1 - 2 265
AK313014.1 - 1 646
BC002771.2 - 2 290
BC063405.1 - 2 290 5190565
BF970430.1 - 511
CA389141.1 - 2 310
CB992246.1 - 2 473

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-vc5lg:80/100.66.78.61:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0