This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HSD17B14 SIRNA Assays ›
Gene Symbol:
HSD17B14
Gene Name:
hydroxysteroid 17-beta dehydrogenase 14
Gene Aliases:
DHRS10, SDR47C1, retSDR3
Chromosome Location:
Chr.19: 48813014 - 48836688 on Build GRCh38
UniGene ID:
Hs.18788
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS181903
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HSD17B14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016246.2 NP_057330.2 3, 4 472
XM_005258969.3 XP_005259026.1 3, 4 481
XM_005258970.3 XP_005259027.1 3, 4 483
GenBank mRNA AF126781.1 - 3, 4 318
AK307437.1 - 3, 4 281
AY358430.1 - 3, 4 311
BC006283.2 - 3, 4 265 4026397
BC006294.1 - 3, 4 273 4025809
BI602824.1 - 3, 4 479

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

estrogen biosynthetic process
steroid catabolic process
oxidation-reduction process
estradiol 17-beta-dehydrogenase activity
protein binding
identical protein binding
testosterone 17-beta-dehydrogenase (NADP+) activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline