This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RNF181 SIRNA Assays ›
Gene Symbol:
RNF181
Gene Name:
ring finger protein 181
Gene Aliases:
HSPC238
Chromosome Location:
Chr.2: 85595268 - 85597708 on Build GRCh38
UniGene ID:
Hs.356187
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS181952
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF181
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016494.3 NP_057578.1 5 493
XM_005264359.3 XP_005264416.1 4 980
GenBank mRNA AF151072.1 - 5 473
AK222700.1 - 5 459
BC002803.2 - 5 468 3636703
CB131242.1 - 5 509
CR457165.1 - 5 443

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001007647, FQ230795, BC079313, FQ213488.

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
protein autoubiquitination
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-tfxwv:80/100.66.74.196:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0