This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IP6K2 SIRNA Assays ›
Gene Symbol:
IP6K2
Gene Name:
inositol hexakisphosphate kinase 2
Gene Aliases:
IHPK2, PIUS
Chromosome Location:
Chr.3: 48688003 - 48717278 on Build GRCh38
UniGene ID:
Hs.595983
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS182053
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IP6K2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005909.2 NP_001005909.1 2 344
NM_001005910.2 NP_001005910.1 2 347
NM_001005911.2 NP_001005911.1 2 344
NM_001146178.2 NP_001139650.1 2 347
NM_001146179.2 NP_001139651.1 2 344
NM_001190316.1 NP_001177245.1 3 560
NM_001190317.1 NP_001177246.1 3 483
NM_016291.3 NP_057375.2 2 347
NR_027437.2 2 347
NR_027438.2 2 344
XM_006713199.3 XP_006713262.1 3 451
XM_006713200.1 XP_006713263.1 3 421
XM_006713201.1 XP_006713264.1 3 498
XM_006713202.1 XP_006713265.1 3 500
XM_011533816.1 XP_011532118.1 3 481
XM_011533817.1 XP_011532119.1 2 284
XM_011533818.1 XP_011532120.1 2 287
XM_011533822.1 XP_011532124.1 2 284
XM_011533823.1 XP_011532125.1 2 287
XM_017006583.1 XP_016862072.1 2 343
XM_017006584.1 XP_016862073.1 2 318
XM_017006585.1 XP_016862074.1 3 433
XM_017006586.1 XP_016862075.1 3 474
XM_017006587.1 XP_016862076.1 3 474
XM_017006588.1 XP_016862077.1 3 500
XM_017006589.1 XP_016862078.1 3 449
XM_017006590.1 XP_016862079.1 3 1055
XM_017006591.1 XP_016862080.1 3 420
XM_017006592.1 XP_016862081.1 3 449
XM_017006593.1 XP_016862082.1 3 449
GenBank mRNA AF177145.1 - 1 241
AK024045.1 - 2 284
AK097016.1 - 8 1202
AK290526.1 - 3 498
AK291501.1 - 3 475
AK304714.1 - 3 421
AK315204.1 - 2 285
AL137514.1 - 2 285
AY007091.1 - 2 270
BC001864.1 - 2 269
BC004469.2 - 2 274
BC019694.1 - 2 256 4941646
BC065533.1 - 2 273 6162048
BP262706.1 - 2 284
BX537544.1 - 3 499
CA488567.1 - 2 281
CD248129.1 - 2 310
CF139656.1 - 2 278
CN362780.1 - 2 259
DA083858.1 - 2 280
DB039955.1 - 2 344

Back To Top

More Information


Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

phosphate ion transport
negative regulation of cell growth
positive regulation of apoptotic process
inositol phosphate metabolic process
phosphatidylinositol phosphorylation
type I interferon signaling pathway
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
protein binding
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-tpmrp:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline