This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
See other IP6K2 SIRNA Assays ›
Gene Symbol:
IP6K2
Gene Name:
inositol hexakisphosphate kinase 2
Gene Aliases:
IHPK2, PIUS
Chromosome Location:
Chr.3: 48688003 - 48717278 on Build GRCh38
UniGene ID:
Hs.595983
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS182054
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IP6K2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005909.2
NP_001005909.1
2
387
NM_001005910.2
NP_001005910.1
2
390
NM_001005911.2
NP_001005911.1
2
387
NM_001146178.2
NP_001139650.1
2
390
NM_001146179.2
NP_001139651.1
2
387
NM_001190316.1
NP_001177245.1
3
603
NM_001190317.1
NP_001177246.1
3
526
NM_016291.3
NP_057375.2
2
390
NR_027437.2
2
390
NR_027438.2
2
387
XM_006713199.3
XP_006713262.1
3
494
XM_006713200.1
XP_006713263.1
3
464
XM_006713201.1
XP_006713264.1
3
541
XM_006713202.1
XP_006713265.1
3
543
XM_011533816.1
XP_011532118.1
3
524
XM_011533817.1
XP_011532119.1
2
327
XM_011533818.1
XP_011532120.1
2
330
XM_011533822.1
XP_011532124.1
2
327
XM_011533823.1
XP_011532125.1
2
330
XM_017006583.1
XP_016862072.1
2
386
XM_017006584.1
XP_016862073.1
2
361
XM_017006585.1
XP_016862074.1
3
476
XM_017006586.1
XP_016862075.1
3
517
XM_017006587.1
XP_016862076.1
3
517
XM_017006588.1
XP_016862077.1
3
543
XM_017006589.1
XP_016862078.1
3
492
XM_017006590.1
XP_016862079.1
3
1098
XM_017006591.1
XP_016862080.1
3
463
XM_017006592.1
XP_016862081.1
3
492
XM_017006593.1
XP_016862082.1
3
492
GenBank mRNA
AF177145.1
-
1
284
AK024045.1
-
2
327
AK097016.1
-
8
1245
AK290526.1
-
3
541
AK291501.1
-
3
518
AK304714.1
-
3
464
AK315204.1
-
2
328
AL137514.1
-
2
328
AY007091.1
-
2
313
BC001864.1
-
2
312
BC004469.2
-
2
317
BC019694.1
-
2
299
4941646
BC065533.1
-
2
316
6162048
BP262706.1
-
2
327
BX537544.1
-
3
542
CA488567.1
-
2
324
CD248129.1
-
2
353
CF139656.1
-
2
321
CN362780.1
-
2
302
DA083858.1
-
2
323
DB039955.1
-
2
387
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
phosphate ion transport
negative regulation of cell growth
positive regulation of apoptotic process
inositol phosphate metabolic process
phosphatidylinositol phosphorylation
type I interferon signaling pathway
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
protein binding
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline