This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LRRN3 SIRNA Assays ›
Gene Symbol:
LRRN3
Gene Name:
leucine rich repeat neuronal 3
Gene Aliases:
FIGLER5, NLRR-3, NLRR3
Chromosome Location:
Chr.7: 111091006 - 111125454 on Build GRCh38
UniGene ID:
Hs.3781
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS182606
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRN3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099658.1 NP_001093128.1 3 1422
NM_001099660.1 NP_001093130.1 4 1529
NM_018334.4 NP_060804.3 2 1340
GenBank mRNA AB060967.1 - 1 789
AF134481.1 - 1 820
AK001991.1 - 4 1441
AK292556.1 - 2 929
AL442092.1 - 1 745
AY358280.1 - 3 962
BC035133.2 - 2 1219 5263900
CR457397.1 - 1 483

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-nrl7w:80/100.66.79.10:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0