This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RBM23 SIRNA Assays ›
Gene Symbol:
RBM23
Gene Name:
RNA binding motif protein 23
Gene Aliases:
CAPERbeta, PP239, RNPC4
Chromosome Location:
Chr.14: 22900645 - 22919187 on Build GRCh38
UniGene ID:
Hs.4997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS182881
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RBM23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077351.1 NP_001070819.1 8 896
NM_001077352.1 NP_001070820.1 6 794
NM_001308044.1 NP_001294973.1 6 735
NM_018107.4 NP_060577.3 7 848
XM_011536890.2 XP_011535192.1 9 979
XM_011536892.2 XP_011535194.1 8 930
XM_011536893.2 XP_011535195.1 8 931
XM_011536894.2 XP_011535196.1 8 925
XM_011536895.2 XP_011535197.1 7 901
XM_011536896.2 XP_011535198.1 7 877
XM_011536897.2 XP_011535199.1 7 1263
XM_011536900.1 XP_011535202.1 7 839
XM_011536902.1 XP_011535204.1 7 745
XM_011536903.1 XP_011535205.1 5 621
XM_011536904.1 XP_011535206.1 6 678
XM_011536905.1 XP_011535207.1 5 676
XM_011536906.1 XP_011535208.1 4 567
XM_017021398.1 XP_016876887.1 8 933
XM_017021399.1 XP_016876888.1 7 880
XM_017021400.1 XP_016876889.1 7 798
XM_017021401.1 XP_016876890.1 8 893
XM_017021402.1 XP_016876891.1 8 804
XM_017021403.1 XP_016876892.1 7 822
XM_017021404.1 XP_016876893.1 7 772
XM_017021405.1 XP_016876894.1 7 839
XM_017021406.1 XP_016876895.1 8 1311
XM_017021407.1 XP_016876896.1 8 820
XM_017021408.1 XP_016876897.1 6 756
XM_017021409.1 XP_016876898.1 7 1257
XM_017021410.1 XP_016876899.1 6 778
XM_017021411.1 XP_016876900.1 6 1209
XM_017021412.1 XP_016876901.1 8 931
XM_017021413.1 XP_016876902.1 7 707
XM_017021414.1 XP_016876903.1 7 745
XM_017021415.1 XP_016876904.1 6 726
XM_017021416.1 XP_016876905.1 5 621
XM_017021417.1 XP_016876906.1 6 678
XM_017021418.1 XP_016876907.1 5 676
XM_017021419.1 XP_016876908.1 4 567
XM_017021420.1 XP_016876909.1 5 1096
GenBank mRNA AF087905.1 - 7 799
AF275678.1 - 8 851
AK001344.1 - 6 744
AK125593.1 - 8 858
AK225792.1 - 8 858
AK303668.1 - 6 697
AK308055.1 - 7 811
AK315620.1 - 7 810
AL834198.1 - 8 857
BC002566.2 - 7 826 3138909
BC024208.2 - 7 785 2960467
BC106012.1 - 5 570
BX161440.1 - 8 829
CR457257.1 - 6 649
KU178687.1 - 6 649
KU178688.1 - 7 697
KU178689.1 - 5 595

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA splicing factor translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
nucleotide binding
RNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-mjcrx:80/100.66.74.196:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0