siRNA Details
| Gene Symbol: LRRC40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017768.4 | NP_060238.3 | 4 | 652 | ||
| XM_005271013.1 | XP_005271070.1 | 3 | 479 | |||
| XM_011541763.1 | XP_011540065.1 | 2 | 394 | |||
| XM_017001695.1 | XP_016857184.1 | 4 | 524 | |||
| XM_017001696.1 | XP_016857185.1 | 2 | 298 | |||
| GenBank mRNA | AK000338.1 | - | 2 | 162 | ||
| AK022951.1 | - | 4 | 566 | |||
| AK225337.1 | - | 2 | 162 | |||
| BC003407.1 | - | 4 | 452 | |||
| BC008586.1 | - | 4 | 550 | 2900193 | ||

LRRC40
Genomic Map