This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C21ORF62 SIRNA Assays ›
Gene Symbol:
C21orf62
Gene Name:
chromosome 21 open reading frame 62
Gene Aliases:
B37, C21orf120, PRED81
Chromosome Location:
Chr.21: 32790673 - 32813808 on Build GRCh38
UniGene ID:
Hs.517235
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS183465
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C21orf62
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162495.2 NP_001155967.2 4 420
NM_001162496.2 NP_001155968.2 2 264
NM_019596.5 NP_062542.5 3 345
GenBank mRNA AF231922.1 - 3 251
AK290983.1 - 3 288
BP214434.1 - 3 305
CN428551.1 - 3 291
DQ656042.2 - 3 345

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline