This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PTMS SIRNA Assays ›
Gene Symbol:
PTMS
Gene Name:
parathymosin
Gene Aliases:
ParaT
Chromosome Location:
Chr.12: 6766139 - 6770952 on Build GRCh38
UniGene ID:
Hs.504613
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS183983
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PTMS
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002824.4 NP_002815.3 2 421
XM_011520987.1 XP_011519289.1 2 657
GenBank mRNA BC017025.1 - 2 140 3900998
BC128229.1 - 3 88
BM550529.1 - 421
M24398.1 - 2 390

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune system process
DNA replication
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0