This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF862 SIRNA Assays ›
Gene Symbol:
ZNF862
Gene Name:
zinc finger protein 862
Gene Aliases:
-
Chromosome Location:
Chr.7: 149838420 - 149867479 on Build GRCh38
UniGene ID:
Hs.731923
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS184900
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF862
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099220.1 NP_001092690.1 4 1085
XM_006716085.3 XP_006716148.1 5 1106
XM_011516463.1 XP_011514765.1 4 1966
XM_011516464.1 XP_011514766.1 4 2355
XM_017012519.1 XP_016868008.1 4 1095
GenBank mRNA AB011115.1 - 2 720

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
molecular_function
nucleic acid binding
metal ion binding
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0