siRNA Details
| Gene Symbol: LOC102723895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120585.1 | 3, 4 | 197 | |||
| GenBank mRNA | DA109127.1 | - | 3, 4 | 197 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723895| Gene Symbol: LOC102723895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120585.1 | 3, 4 | 197 | |||
| GenBank mRNA | DA109127.1 | - | 3, 4 | 197 | ||