This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SMIM23 SIRNA Assays ›
Gene Symbol:
SMIM23
Gene Name:
small integral membrane protein 23
Gene Aliases:
C5orf50
Chromosome Location:
Chr.5: 171772637 - 171791136 on Build GRCh38
UniGene ID:
Hs.591740
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS185122
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SMIM23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289970.1 NP_001276899.1 3 188
XM_011534622.1 XP_011532924.1 3 271
XM_011534623.2 XP_011532925.1 3 382
XM_011534624.2 XP_011532926.1 4 270
GenBank mRNA EG327678.1 - 3 133

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-khs54:80/100.66.77.55:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0