This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM227A SIRNA Assays ›
Gene Symbol:
FAM227A
Gene Name:
family with sequence similarity 227 member A
Gene Aliases:
-
Chromosome Location:
Chr.22: 38578120 - 38656629 on Build GRCh38
UniGene ID:
Hs.542707
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS185396
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM227A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013647.1 NP_001013669.1 5 920
NM_001291030.1 NP_001277959.1 5 960
XM_006724291.2 XP_006724354.2 5 920
XM_006724292.2 XP_006724355.2 5 920
XM_006724293.2 XP_006724356.1 5 920
XM_006724294.2 XP_006724357.2 5 960
XM_006724295.2 XP_006724358.2 5 960
XM_011530308.2 XP_011528610.1 5 625
XM_011530309.2 XP_011528611.1 5 920
XM_011530310.1 XP_011528612.1 5 920
XM_011530311.1 XP_011528613.1 4 797
XM_011530312.1 XP_011528614.1 3 197
XM_011530313.2 XP_011528615.1 3 149
XM_011530314.1 XP_011528616.1 4 850
GenBank mRNA AK301819.1 - 5 634
DB053353.1 - 5 549

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0