This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BMX SIRNA Assays ›
Gene Symbol:
BMX
Gene Name:
BMX non-receptor tyrosine kinase
Gene Aliases:
ETK, PSCTK2, PSCTK3
Chromosome Location:
Chr.X: 15500777 - 15556529 on Build GRCh38
UniGene ID:
Hs.495731
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS185998
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BMX
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320866.1 NP_001307795.1 4 444
NM_001721.6 NP_001712.1 4 382
NM_203281.2 NP_975010.1 4 444
XM_017029752.1 XP_016885241.1 4 414
GenBank mRNA AF045459.1 - 4 458
AK291824.1 - 4 414
AK313568.1 - 4 377
BC016652.2 - 4 399 3919253
BP236451.1 - 4 444
BQ226991.1 - 4 404
CB998198.1 - 419
X83107.1 - 4 304

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

NK T cell differentiation
protein phosphorylation
cellular component disassembly involved in execution phase of apoptosis
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
mesoderm development
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
innate immune response
protein autophosphorylation
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
signal transducer activity
receptor binding
protein binding
ATP binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-jq955:80/100.66.72.86:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0