This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF419 SIRNA Assays ›
Gene Symbol:
ZNF419
Gene Name:
zinc finger protein 419
Gene Aliases:
ZAPHIR, ZNF419A
Chromosome Location:
Chr.19: 57487711 - 57494680 on Build GRCh38
UniGene ID:
Hs.744339
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS188158
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF419
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098491.1 NP_001091961.1 5 1631
NM_001098492.1 NP_001091962.1 4 1592
NM_001098493.1 NP_001091963.1 4 1589
NM_001098494.1 NP_001091964.1 4 1532
NM_001098495.1 NP_001091965.1 3 1493
NM_001098496.1 NP_001091966.1 3 1490
NM_001291743.1 NP_001278672.1 4 1529
NM_001291744.1 NP_001278673.1 4 1548
NM_024691.3 NP_078967.3 5 1628
GenBank mRNA AB209371.1 - 3 4877
AK026886.1 - 4 1502
AK302135.1 - 3 1451
AK304569.1 - 4 1537
AK309427.1 - 3 2095
AK316156.1 - 4 1550
AK316297.1 - 4 1582
BC008297.1 - 5 1574 3544044
BC024259.1 - 5 1574 3604215

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline