This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF419 SIRNA Assays ›
Gene Symbol:
ZNF419
Gene Name:
zinc finger protein 419
Gene Aliases:
ZAPHIR, ZNF419A
Chromosome Location:
Chr.19: 57487711 - 57494680 on Build GRCh38
UniGene ID:
Hs.744339
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS188159
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF419
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098491.1 NP_001091961.1 5 612
NM_001098492.1 NP_001091962.1 4 573
NM_001098493.1 NP_001091963.1 4 570
NM_001098494.1 NP_001091964.1 4 513
NM_001098495.1 NP_001091965.1 3 474
NM_001098496.1 NP_001091966.1 3 471
NM_001291743.1 NP_001278672.1 4 510
NM_001291744.1 NP_001278673.1 4 529
NM_024691.3 NP_078967.3 5 609
GenBank mRNA AB209371.1 - 3 3858
AK026886.1 - 4 483
AK302135.1 - 3 432
AK304569.1 - 4 518
AK307802.1 - 5 601
AK309427.1 - 3 1076
AK316156.1 - 4 531
AK316297.1 - 4 563
BC008297.1 - 5 555 3544044
BC024259.1 - 5 555 3604215
CT002615.1 - 4 440
DA548110.1 - 4 526
DC407547.1 - 3 432
DC425209.1 - 3 442

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline