siRNA Details
| Gene Symbol: ZNF671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321375.1 | NP_001308304.1 | 3 | 1031 | ||
| NM_001321376.1 | NP_001308305.1 | 5 | 1196 | |||
| NM_024833.2 | NP_079109.2 | 4 | 1161 | |||
| XM_017027314.1 | XP_016882803.1 | 4 | 1654 | |||
| GenBank mRNA | AK027159.1 | - | 4 | 1149 | ||
| AK091421.1 | - | 5 | 1196 | |||
| AK225665.1 | - | 4 | 1158 | |||
| AK293680.1 | - | 3 | 1029 | |||
| BC025728.1 | - | 4 | 1108 | 5202908 | ||

ZNF671
Genomic Map